Merck
  • Increased expression of the transforming growth factor β-inducible gene HIC-5 in systemic sclerosis skin and fibroblasts: a novel antifibrotic therapeutic target.
Increased expression of the transforming growth factor β-inducible gene HIC-5 in systemic sclerosis skin and fibroblasts: a novel antifibrotic therapeutic target.
Rheumatology (Oxford, England) (2020-05-23)
Sonsoles Piera-Velazquez, Jolanta Fertala, Gonzalo Huaman-Vargas, Natalia Louneva, Sergio A Jiménez
RESUMEN
SSc is a systemic fibrotic disease affecting skin, numerous internal organs and the microvasculature. The molecular pathogenesis of SSc tissue fibrosis has not been fully elucidated, although TGF-β1 plays a crucial role. The Hic-5 protein encoded by the TGF-β1-inducible HIC-5 gene participates in numerous TGF-β-mediated pathways, however, the role of Hic-5 in SSc fibrosis has not been investigated. The aim of this study was to examine HIC-5 involvement in SSc tissue fibrosis. Affected skin from three patients with diffuse SSc and dermal fibroblasts cultured from affected and non-affected SSc skin were examined for HIC-5 and COL1A1 gene expression. Real-time PCR, IF microscopy, western blotting and small interfering RNA-mediated HIC-5 were performed. HIC-5 and COL1A1 transcripts and Hic-5, type 1 collagen (COL1) and α-smooth muscle actin (α-SMA) protein levels were increased in clinically affected SSc skin compared with normal skin and in cultured dermal fibroblasts from affected SSc skin compared with non-affected skin fibroblasts from the same patients. HIC-5 knockdown caused a marked reduction of COL1 production in SSc dermal fibroblasts. HIC-5 expression is increased in affected SSc skin compared with skin from normal individuals. Affected SSc skin fibroblasts display increased HIC-5 and COL1A1 expression compared with non-affected skin fibroblasts from the same patients. Hic-5 protein was significantly increased in cultured SSc dermal fibroblasts. HIC-5 mRNA knockdown in SSc fibroblasts caused >50% reduction of COL1 production. Although these are preliminary results owing to the small number of skin samples studied, they indicate that Hic-5 plays a role in the profibrotic activation of SSc dermal fibroblasts and may represent a novel molecular target for antifibrotic therapy in SSc.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting human TGFB1I1