Merck
Structure-activity relationships of 7-deaza-6-benzylthioinosine analogues as ligands of Toxoplasma gondii adenosine kinase.
Journal of medicinal chemistry (2008-06-20)
Young Ah Kim, Ashoke Sharon, Chung K Chu, Reem H Rais, Omar N Al Safarjalani, Fardos N M Naguib, Mahmoud H el Kouni
RESUMEN
Several 7-deaza-6-benzylthioinosine analogues with varied substituents on aromatic ring were synthesized and evaluated against Toxoplasma gondii adenosine kinase (EC.2.7.1.20). Structure-activity relationships indicated that the nitrogen atom at the 7-position does not appear to be a critical structural requirement. Molecular modeling reveals that the 7-deazapurine motif provided flexibility to the 6-benzylthio group as a result of the absence of H-bonding between N7 and Thr140. This flexibility allowed better fitting of the 6-benzylthio group into the hydrophobic pocket of the enzyme at the 6-position. In general, single substitutions at the para or meta position enhanced binding. On the other hand, single substitutions at the ortho position led to the loss of binding affinity. The most potent compounds, 7-deaza- p-cyano-6-benzylthioinosine (IC 50 = 5.3 microM) and 7-deaza- p-methoxy-6-benzylthioinosine (IC 50 = 4.6 microM), were evaluated in cell culture to delineate their selective toxicity.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
Adenosine, BioReagent, suitable for cell culture
Sigma-Aldrich
Sulfadiazine, 99.0-101.0%
Supelco
Sulfadiazine, VETRANAL®, analytical standard