Merck
Enrichment of the tumour immune microenvironment in patients with desmoplastic colorectal liver metastasis.
British journal of cancer (2020-05-19)
Diederik J Höppener, Pieter M H Nierop, Joost Hof, Kostandinos Sideras, Guoying Zhou, Lydia Visser, Annette S H Gouw, Koert P de Jong, Dave Sprengers, Jaap Kwekkeboom, Peter B Vermeulen, Dirk J Grünhagen, Cornelis Verhoef
RESUMEN
Patients with resected colorectal liver metastasis (CRLM) who display only the desmoplastic histopathological growth pattern (dHGP) exhibit superior survival compared to patients with any non-desmoplastic growth (non-dHGP). The aim of this study was to compare the tumour microenvironment between dHGP and non-dHGP. The tumour microenvironment was investigated in three cohorts of chemo-naive patients surgically treated for CRLM. In cohort A semi-quantitative immunohistochemistry was performed, in cohort B intratumoural and peritumoural T cells were counted using immunohistochemistry and digital image analysis, and in cohort C the relative proportions of individual T cell subsets were determined by flow cytometry. One hundred and seventeen, 34, and 79 patients were included in cohorts A, B, and C, with dHGP being observed in 27%, 29%, and 15% of patients, respectively. Cohorts A and B independently demonstrated peritumoural and intratumoural enrichment of cytotoxic CD8+ T cells in dHGP, as well as a higher CD8+/CD4+ ratio (cohort A). Flow cytometric analysis of fresh tumour tissues in cohort C confirmed these results; dHGP was associated with higher CD8+ and lower CD4+ T cell subsets, resulting in a higher CD8+/CD4+ ratio. The tumour microenvironment of patients with dHGP is characterised by an increased and distinctly cytotoxic immune infiltrate, providing a potential explanation for their superior survival.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-SLAMF7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
En este momento no podemos mostrarle ni los precios ni la disponibilidad