Merck
Role of vimentin in modulating immune cell apoptosis and inflammatory responses in sepsis.
Scientific reports (2019-04-07)
Longxiang Su, Pan Pan, Peng Yan, Yun Long, Xiang Zhou, Xiaoting Wang, Ruo Zhou, Bo Wen, Lixin Xie, Dawei Liu
RESUMEN
New diagnostic biomarkers or therapeutic targets for sepsis have substantial significance for critical care medicine. In this study, 192 differentially expressed proteins were selected through iTRAQ. Based on cluster analysis of protein expression dynamics and protein-protein interactions, hemopexin, vimentin, and heat shock protein 90 were selected for further investigation. It was demonstrated that serum vimentin (VIM) levels were significantly increased in patients with sepsis and septic shock compared to controls and that VIM expression was significantly increased in lymphocytes isolated from septic shock and sepsis patients compared to controls. Moreover, a nonsurvivor group had higher serum VIM levels and VIM expression in lymphocytes. Caspase-3 was significantly upregulated in Jurkat T cells lacking VIM and when exposed to LPS compared to control cells. In contrast, caspase-3 was reduced nearly 40% in cells over-expressing VIM. IL-2, IL-10 and IFN-α levels were significantly decreased in cells lacking VIM compared to control cells, whereas they were not significantly altered in cells over-expressing VIM. These findings suggest that VIM modulates lymphocyte apoptosis and inflammatory responses and that VIM could be a new target for the diagnosis and prognostic prediction of patients with sepsis or septic shock.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Vimentin, His tagged human, recombinant, expressed in E. coli, ≥90% (SDS-PAGE), buffered aqueous glycerol solution