Merck
C-geranylated chalcones from the stems of Angelica keiskei with superoxide-scavenging activity.
Journal of natural products (2008-06-19)
Nobuwa Aoki, Mai Muko, Emi Ohta, Shinji Ohta
RESUMEN
An ethyl acetate-soluble fraction of a methanol extract from the stems of Angelica keiskei was subjected to chromatographic separation to give three new chalcones, designated as xanthokeismins A-C (1-3), in addition to a known chalcone, xanthoangelol B (4). The structures of 1-3 were determined on the basis of the analysis of their spectroscopic data. Compounds 1-4 exhibited potent superoxide-scavenging activity.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Ácido L-ascórbico, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Ácido L-ascórbico, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Ácido L-ascórbico, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Ácido L-ascórbico, 99%
Sigma-Aldrich
Ácido L-ascórbico, reagent grade, crystalline
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
Ácido L-ascórbico, ACS reagent, ≥99%
Supelco
Ácido L-ascórbico, analytical standard
Sigma-Aldrich
Ácido L-ascórbico, meets USP testing specifications
Sigma-Aldrich
Ácido L-ascórbico, reagent grade
Sigma-Aldrich
Farnesol, 95%
Sigma-Aldrich
Ácido L-ascórbico, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Ácido L-ascórbico, FCC, FG
Sigma-Aldrich
Ácido L-ascórbico, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
trans,trans-Farnesol, 96%
Sigma-Aldrich
Ácido L-ascórbico, puriss. p.a., ≥99.0% (RT)
Supelco
Resveratrol, analytical standard
Sigma-Aldrich
Farnesol, mixture of isomers, ≥95%, stabilized, FG
Sigma-Aldrich
Ácido L-ascórbico, tested according to Ph. Eur.