Merck
Comparing anti-HIV, antibacterial, antifungal, micellar, and cytotoxic properties of tricarboxylato dendritic amphiphiles.
Bioorganic & medicinal chemistry (2009-03-25)
Richard V Macri, Janka Karlovská, Gustavo F Doncel, Xiaosong Du, Bhadreshkumar B Maisuria, André A Williams, Eko W Sugandhi, Joseph O Falkinham, Alan R Esker, Richard D Gandour
RESUMEN
Three series of homologous dendritic amphiphiles--RCONHC(CH(2)CH(2)COOH)(3), 1(n); ROCONHC(CH(2)CH(2)COOH)(3), 2(n); RNHCONHC(CH(2)CH(2)COOH)(3), 3(n), where R = n-C(n)H(2n+1) and n = 13-22 carbon atoms--were assayed for their potential to serve as antimicrobial components in a topical vaginal formulation. Comparing epithelial cytotoxicities to the ability of these homologues to inhibit HIV, Neisseria gonorrhoeae, and Candida albicans provided a measure of their prophylactic/therapeutic potential. Measurements of the ability to inhibit Lactobacillus plantarum, a beneficial bacterium in the vagina, and critical micelle concentrations (CMCs), an indicator of the potential detergency of these amphiphiles, provided additional assessments of safety. Several amphiphiles from each homologous series had modest anti-HIV activity (EC(50) = 110-130 microM). Amphiphile 2(18) had the best anti-Neisseria activity (MIC =65 microM), while 1(19) and 1(21) had MICs against C. albicans of 16 and 7.7 microM, respectively. Two measures of safety showed promise as all compounds had relatively low cytotoxic activity (EC(50) = 210-940 microM) against epithelial cells and low activity against L. plantarum, 1(n), 2(n), and 3(n) had MICs490, 1300, and 940 microM, respectively. CMCs measured in aqueous triethanolamine and in aqueous potassium hydroxide showed linear dependences on chain length. As expected, the longest chain in each series had the lowest CMC-in triethanolamine: 1(21), 1500 microM; 2(22), 320 microM; 3(22), 340 microM, and in potassium hydroxide: 1(21), 130 microM; 3(22), 40 microM. The CMC in triethanolamine adjusted to pH 7.4 was 400 microM for 1(21) and 3900 microM for 3(16). The promising antifungal activity, low activity against L. plantarum, relatively high CMCs, and modest epithelial cytotoxicity in addition to their anti-Neisseria properties warrant further design studies with dendritic amphiphiles to improve their safety indices to produce suitable candidates for antimicrobial vaginal products.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Trietanolamina, ≥99.0% (GC)
Sigma-Aldrich
Trietanolamina, reagent grade, 98%
Sigma-Aldrich
Trietanolamina, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Trietanolamina, puriss. p.a., ≥99% (GC)
Sigma-Aldrich
Trietanolamina, puriss., meets analytical specification of NF, ≥99% (GC)