Merck
  • FGF-2 Stimulates the Growth of Tenogenic Progenitor Cells to Facilitate the Generation of Tenomodulin-Positive Tenocytes in a Rat Rotator Cuff Healing Model.
FGF-2 Stimulates the Growth of Tenogenic Progenitor Cells to Facilitate the Generation of Tenomodulin-Positive Tenocytes in a Rat Rotator Cuff Healing Model.
The American journal of sports medicine (2015-08-28)
Takuya Tokunaga, Chisa Shukunami, Nobukazu Okamoto, Takuya Taniwaki, Kiyoshi Oka, Hidetoshi Sakamoto, Junji Ide, Hiroshi Mizuta, Yuji Hiraki
RESUMEN
Fibroblast growth factor (FGF)-2 has the potential to enhance tendon-to-bone healing after rotator cuff (RC) injury. FGF-2 stimulates tenogenic differentiation of progenitors to improve the biomechanical strength and histological appearance of repaired RCs in rats. Controlled laboratory study. Adult male Sprague-Dawley rats (N = 156) underwent unilateral surgery to repair the supraspinatus tendon to insertion sites. The FGF-2-treated group (gelatin hydrogel containing 5 μg of FGF-2) and a control group (gelatin hydrogel only) were compared to investigate the effects of FGF-2 at 2, 4, 6, 8, and 12 weeks postoperatively. Biomechanical testing was performed at 6 and 12 weeks. Semiquantitative histological analysis and immunohistochemical analysis for the proliferating cell nuclear antigen (PCNA) were performed, and the expression of tendon-related markers, including Scleraxis (Scx) and Tenomodulin (Tnmd), was monitored by real-time reverse transcription polymerase chain reaction (RT-PCR) and in situ hybridization. SRY-box containing gene 9 (Sox9) expression was monitored by RT-PCR and immunohistochemical analysis. At 2 and 4 weeks, immunohistochemical analysis for mesenchymal stem cell (MSC) markers was also performed. The FGF-2-treated group demonstrated a significant improvement in mechanical strength at 6 and 12 weeks and significantly higher histological scores than the control group at ≥4 weeks. The average incidence of PCNA-positive cells was significantly higher at 2 and 4 weeks, and more cells expressing MSC markers were detected at the insertion site in the FGF-2-treated group. The expression level of Scx increased significantly in the FGF-2-treated group from 4 to 8 weeks, while the Tnmd level increased significantly from 4 to 12 weeks postoperatively. The localization of Tnmd overlapped with the locations of reparative tissues accompanying collagen fibers with an aligned orientation. Sox9 expression was significantly upregulated at 4 weeks in the FGF-2-treated group. FGF-2 promotes growth of the tenogenic progenitor cells, which participate in tendon-to-bone healing, resulting in biomechanical and histological improvement of the repaired RC. These findings provide clues regarding the clinical development of regenerative repair strategies for RC injury.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Anticuerpo anti-Sox9, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Endoglin Antibody, clone P3D1, clone P3D1, from mouse