Merck
  • An Autochthonous Mouse Model of Myd88- and BCL2-Driven Diffuse Large B-cell Lymphoma Reveals Actionable Molecular Vulnerabilities.
An Autochthonous Mouse Model of Myd88- and BCL2-Driven Diffuse Large B-cell Lymphoma Reveals Actionable Molecular Vulnerabilities.
Blood cancer discovery (2021-01-16)
Ruth Flümann, Tim Rehkämper, Pascal Nieper, Pauline Pfeiffer, Alessandra Holzem, Sebastian Klein, Sanil Bhatia, Moritz Kochanek, Ilmars Kisis, Benedikt W Pelzer, Heinz Ahlert, Julia Hauer, Alexandra da Palma Guerreiro, Jeremy A Ryan, Maurice Reimann, Arina Riabinska, Janica Wiederstein, Marcus Krüger, Martina Deckert, Janine Altmüller, Andreas R Klatt, Lukas P Frenzel, Laura Pasqualucci, Wendy Béguelin, Ari M Melnick, Sandrine Sander, Manuel Montesinos-Rongen, Anna Brunn, Philipp Lohneis, Reinhard Büttner, Hamid Kashkar, Arndt Borkhardt, Anthony Letai, Thorsten Persigehl, Martin Peifer, Clemens A Schmitt, Hans Christian Reinhardt, Gero Knittel
RESUMEN
Based on gene expression profiles, diffuse large B cell lymphoma (DLBCL) is sub-divided into germinal center B cell-like (GCB) and activated B cell-like (ABC) DLBCL. Two of the most common genomic aberrations in ABC-DLBCL are mutations in MYD88, as well as BCL2 copy number gains. Here, we employ immune phenotyping, RNA-Seq and whole exome sequencing to characterize a Myd88 and Bcl2-driven mouse model of ABC-DLBCL. We show that this model resembles features of human ABC-DLBCL. We further demonstrate an actionable dependence of our murine ABC-DLBCL model on BCL2. This BCL2 dependence was also detectable in human ABC-DLBCL cell lines. Moreover, human ABC-DLBCLs displayed increased PD-L1 expression, compared to GCB-DLBCL. In vivo experiments in our ABC-DLBCL model showed that combined venetoclax and RMP1-14 significantly increased the overall survival of lymphoma bearing animals, indicating that this combination may be a viable option for selected human ABC-DLBCL cases harboring MYD88 and BCL2 aberrations.
MATERIALES
Número de producto
Marca
Descripción del producto
Roche
cOmplete ULTRA Tablets, Mini, EASYpack Protease Inhibitor Cocktail, Tablets supplied in foil blister packs.
Sigma-Aldrich
Digitonina, ~50% (TLC)
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS