Merck
Germline transgenesis in rodents by pronuclear microinjection of Sleeping Beauty transposons.
Nature protocols (2014-03-15)
Zoltán Ivics, Lajos Mátés, Tien Yin Yau, Vladimír Landa, Vaclav Zidek, Sanum Bashir, Orsolya I Hoffmann, László Hiripi, Wiebke Garrels, Wilfried A Kues, Zsuzsanna Bösze, Aron Geurts, Michal Pravenec, Thomas Rülicke, Zsuzsanna Izsvák
RESUMEN
We describe a protocol for high-efficiency germline transgenesis and sustained transgene expression in two important biomedical models, the mouse and the rat, by using the Sleeping Beauty transposon system. The procedure is based on co-injection of synthetic mRNA encoding the SB100X hyperactive transposase, together with circular plasmid DNA carrying a transgene construct flanked by binding sites for the transposase, into the pronuclei of fertilized oocytes. Upon translation of the transposase mRNA, enzyme-mediated excision of the transgene cassettes from the injected plasmids followed by permanent genomic insertion produces stable transgenic animals. Generation of a germline-transgenic founder animal by using this protocol takes ∼3 months. Transposon-mediated transgenesis compares favorably in terms of both efficiency and reliable transgene expression with classic pronuclear microinjection, and it offers comparable efficacies to lentiviral approaches without limitations on vector design, issues of transgene silencing, and the toxicity and biosafety concerns of working with viral vectors.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Agua, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Cloruro de sodio, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Dietilpirocarbonato, 96% (NT)
Sigma-Aldrich
Medio M2, With HEPES, without penicillin and streptomycin, liquid, sterile-filtered, suitable for mouse embryo cell culture
Sigma-Aldrich
Tampón Tris-EDTA, BioUltra, Molecular Biology, pH 8.0
Sigma-Aldrich
Penicillin G sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethidium bromide, BioReagent, Molecular Biology, powder
Sigma-Aldrich
M16 Medium, With sodium bicarbonate and lactic acid, without penicillin and streptomycin, liquid, sterile-filtered
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture