Merck
Oxytocin Modulation of Maternal Behavior and Its Association With Immunological Activity in Rats With Cesarean Delivery.
ASN neuro (2021-07-03)
Tong Li, Shu-Wei Jia, Dan Hou, Xiaoran Wang, Dongyang Li, Yang Liu, Dan Cui, Xiaoyu Liu, Chun-Mei Hou, Ping Wang, Colin H Brown, Yu-Feng Wang
RESUMEN
Oxytocin (OT), a neuropeptide produced in the supraoptic (SON) and paraventricular (PVN) nuclei, is not only essential for lactation and maternal behavior but also for normal immunological activity. However, mechanisms underlying OT regulation of maternal behavior and its association with immunity around parturition, particularly under mental and physical stress, remain unclear. Here, we observed effects of OT on maternal behavior in association with immunological activity in rats after cesarean delivery (CD), a model of reproductive stress. CD significantly reduced maternal interests to the pups throughout postpartum day 1-8. On postpartum day 5, CD decreased plasma OT levels and thymic index but increased vasopressin, interleukin (IL)-1β, IL-6 and IL-10 levels. CD had no significant effect on plasma adrenocorticotropic hormone and corticosterone levels. In the hypothalamus, CD decreased corticotropin-releasing hormone contents in the PVN but increased OT contents in the PVN and SON and OT release from hypothalamic implants. CD also increased c-Fos expression, particularly in the cytoplasm of OT neurons. Lastly, CD depolarized resting membrane potential and increased spike width while increasing the variability of the firing rate of OT neurons in brain slices. Thus, CD can increase hypothalamic OT contents and release but reduce pituitary release of OT into the blood, which is associated with depressive-like maternal behavior, increased inflammatory cytokine release and decreased relative weight of the thymus.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Cóctel de inhibidores de proteasas, for use with mammalian cell and tissue extracts, DMSO solution
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Dodecilsulfatosódico, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
L-Glutamina solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Disolución de tripsina-EDTA, 1 ×, sterile; sterile-filtered, BioReagent, suitable for cell culture, 0.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Seroalbúmina bovina, lyophilized powder, BioReagent, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Roche
Kit de detección de muerte celular in situ,TMR rojo, sufficient for ≤50 tests
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Evans Blue, Dye content ≥75 %
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Poli-D-lisina hydrobromide, average mol wt 30,000-70,000, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Biocitina, ≥98% (TLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
SIGMAFAST 3,3′-Diaminobenzidine tablets, tablet, To prepare 5 mL
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Roche
DAPI, 4′,6-Diamidine-2′-phenylindole dihydrochloride
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
Ácido bicinconínico solution
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Poli-L-lisina hydrobromide, mol wt 150,000-300,000
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
bisBenzimide H 33342 trihydrochloride, ≥98% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
ISRIB, ≥98% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Gentamicina solution, Hybri-Max, 50 mg/mL gentamicin, 0.1 μm filtered, BioReagent, suitable for hybridoma
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Glutamine and Glutamate Determination Kit
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Anticuerpo anti-neurofisina 1, clon PS 38, clone PS 38, from mouse
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
LY 294002
En este momento no podemos mostrarle ni los precios ni la disponibilidad