Merck
  • Sera from people with HIV and depression induce commensurate metabolic alterations in astrocytes: toward precision diagnoses and therapies.
Sera from people with HIV and depression induce commensurate metabolic alterations in astrocytes: toward precision diagnoses and therapies.
NeuroImmune pharmacology and therapeutics (2024-08-23)
Anna Elizabeth Laird, Alexandra Anh Le, Jacqueline R Kulbe, Anya Umlauf, Melody Sagarian, Matthew Spencer, Anish Sathe, David J Grelotti, Jennifer Iudicello, Brook Henry, Ronald J Ellis, Jerel Adam Fields
RESUMEN
People with HIV (PWH) have high rates of depression and neurocognitive impairment (NCI) despite viral suppression on antiretroviral therapy (ART). Mounting evidence suggests that immunometabolic disruptions may contribute to these conditions in some PWH. We hypothesized that metabolic dysfunction in astrocytes is associated with depressive symptoms and cognitive function in PWH. Human astrocytes were exposed to sera from PWH (n=40) with varying degrees of depressive symptomatology and cognitive function. MitoTrackerTM Deep Red FM (MT) was used to visualize mitochondrial activity and glial fibrillary acidic protein (GFAP) as an indicator of astrocyte reactivity using the high-throughput fluorescent microscopy and image analyses platform, CellInsight CX5 (CX5). The Seahorse platform was used to assess glycolytic and mitochondrial metabolism. More severe depression, as indexed by higher Beck's Depression Inventory (BDI-II) scores, was associated with lower MT signal measures. Better cognitive function, as assessed by neuropsychiatric testing t-scores, was associated with increased MT signal measures. GFAP intensity negatively correlated with several cognitive t-scores. Age positively correlated with (higher) MT signal measures and GFAP intensity. Worse depressive symptoms (higher BDI-II scores) were associated with decreased oxygen consumption rate and spare respiratory capacity, concomitant with increased extracellular acidification rate in astrocytes. These findings show that factors in the sera of PWH alter mitochondrial activity in cultured human astrocytes, suggesting that mechanisms that alter mitochondrial and astrocyte homeostasis can be detected peripherally. Thus, in vitro cultures may provide a model to identify neuropathogenic mechanisms of depression or neurocognitive impairment in PWH and test personalized therapeutics for neurologic and psychiatric disorders.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Anti-proteína gliofibrilar ácida (GFAP) monoclonal antibody produced in mouse, clone G-A-5, ascites fluid