Merck
Differential contribution of Bacillus anthracis toxins to pathogenicity in two animal models.
Infection and immunity (2012-05-16)
Haim Levy, Shay Weiss, Zeev Altboum, Josef Schlomovitz, Itai Glinert, Assa Sittner, Avigdor Shafferman, David Kobiler
RESUMEN
The virulence of Bacillus anthracis, the causative agent of anthrax, stems from its antiphagocytic capsule, encoded by pXO2, and the tripartite toxins encoded by pXO1. The accepted paradigm states that anthrax is both an invasive and toxinogenic disease and that the toxins play major roles in pathogenicity. We tested this assumption by a systematic study of mutants with combined deletions of the pag, lef, and cya genes, encoding protective antigen (PA), lethal factor (LF), and edema factor (EF), respectively. The resulting seven mutants (single, double, and triple) were evaluated following subcutaneous (s.c.) and intranasal (i.n.) inoculation in rabbits and guinea pigs. In the rabbit model, virulence is completely dependent on the presence of PA. Any mutant bearing a pag deletion behaved like a pXO1-cured mutant, exhibiting complete loss of virulence with attenuation indices of over 2,500,000 or 1,250 in the s.c. or i.n. route of infection, respectively. In marked contrast, in guinea pigs, deletion of pag or even of all three toxin components resulted in relatively moderate attenuation, whereas the pXO1-cured bacteria showed complete attenuation. The results indicate that a pXO1-encoded factor(s), other than the toxins, has a major contribution to the virulence mechanism of B. anthracis in the guinea pig model. These unexpected toxin-dependent and toxin-independent manifestations of pathogenicity in different animal models emphasize the importance and need for a comprehensive evaluation of B. anthracis virulence in general and in particular for the design of relevant next-generation anthrax vaccines.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
AccuTaq LA DNA Polymerase, High fidelity Taq enzyme, with 10X buffer & DMSO