Merck
  • Correction of protein metabolic disorders by composite extract of Musa paradisiaca and Coccinia indica in streptozotocin-induced diabetic albino rat: an approach through the pancreas.
Correction of protein metabolic disorders by composite extract of Musa paradisiaca and Coccinia indica in streptozotocin-induced diabetic albino rat: an approach through the pancreas.
Pancreas (2009-01-27)
Chhanda Mallick, Debasis De, Debidas Ghosh
RESUMEN
The study focused on the ability of the extracts of Musa paradisiaca and Coccinia indica on protein metabolic disorders in streptozotocin (STZ)-induced diabetes. Wistar strain rats were divided into 6 groups as control, control + composite extract treated, STZ-induced diabetes, diabetic + composite extract treated, composite extract-pretreated diabetes, and composite extract-pretreated diabetes + composite extract treated. Protein metabolic status was assessed by serum levels of urea, uric acid, albumin, and creatinine along with urine urea and albumin levels. Diabetic therapeutic ability was assessed by blood glucose, glycated hemoglobin, and serum insulin levels. Histology of the pancreas, liver, and kidney was evaluated. Indices of protein metabolic disorders were deviated from control in STZ-induced diabetes, which were protected significantly after the treatment of composite extract of M. paradisiaca and C. indica. This protection was more prominent when the extract-pretreated animals were subjected to diabetes induction by STZ. The composite extract has a protective therapeutic effect against diabetes through beta-cell regeneration capacity.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Creatininase from microorganisms, lyophilized powder, 100-300 units/mg protein