Merck
  • Dehydroepiandrosterone and dehydroepiandrosterone sulfate: anabolic, neuroprotective, and neuroexcitatory properties in military men.
Dehydroepiandrosterone and dehydroepiandrosterone sulfate: anabolic, neuroprotective, and neuroexcitatory properties in military men.
Military medicine (2013-01-30)
Marcus K Taylor
RESUMEN
Evidence links dehydroepiandrosterone (DHEA) and dehydroepiandrosterone sulfate (DHEAS) to crucial military health issues, including operational stress, resilience, and traumatic brain injury. This study evaluated the anabolic, neuroprotective, and neuroexcitatory properties of DHEA(S) in healthy military men. A salivary sample was obtained from 42 men and assayed for DHEA(S), testosterone, nerve growth factor (NGF; which supports nerve cell proliferation), and salivary alpha amylase (sAA; a proxy of sympathetic nervous system function). Separate regression analyses were conducted with DHEA and DHEAS as independent variables, and testosterone, NGF, and sAA as dependent variables, respectively. The models explained 23.4% of variance in testosterone (p < 0.01), 17.2% of variance in NGF (p < 0.01), and 7.4% of variance in sAA (p = 0.09). Standardized beta coefficients revealed that DHEA independently influenced testosterone (beta = 0.40, p < 0.01), whereas DHEAS independently influenced NGF (beta = 0.48, p < 0.01) and sAA (beta = 0.36, p < 0.05). DHEA demonstrated anabolic properties, whereas DHEAS demonstrated neuroprotective and neuroexcitatory properties in military men. This area of study has broad implications for stress inoculation, traumatic brain injury rehabilitation, and regenerative medicine in military personnel.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
αα-Amilasa termoestable, solution, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
αα-Amilasa from human saliva, Type XIII-A, lyophilized powder, 300-1,500 units/mg protein
Sigma-Aldrich
α-Amylase from porcine pancreas, Type VI-B, ≥5 units/mg solid
Sigma-Aldrich
αα-amilasa from Bacillus sp., Type II-A, lyophilized powder, ≥1,500 units/mg protein (biuret)
Sigma-Aldrich
Taka-diastasa from Aspergillus oryzae, powder, slightly beige, ~100 U/mg
Sigma-Aldrich
αα-Amilasa from Aspergillus oryzae, ≥150 units/mg protein (biuret)
Sigma-Aldrich
αα-Amilasa from Aspergillus oryzae, powder, ~30 U/mg
Sigma-Aldrich
αα-Amilasa from human saliva, Type IX-A, lyophilized powder, 1,000-3,000 units/mg protein
Sigma-Aldrich
αα-Amilasa from Bacillus licheniformis, lyophilized powder, 500-1,500 units/mg protein, 93-100% (SDS-PAGE)
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~50 U/mg
Sigma-Aldrich
αα-Amilasa from Bacillus licheniformis, Type XII-A, saline solution, ≥500 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from porcine pancreas, PMSF Treated, Type I-A, saline suspension, ≥1000 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from Bacillus sp., liquid
Sigma-Aldrich
trans-Dehydroandrosterone, ≥99%
Sigma-Aldrich
αα-Amilasa from Aspergillus oryzae, powder, ~1.5 U/mg (~0.2 U acc. to Willstätter)
Sigma-Aldrich
αα-amilasa from Bacillus sp., powder, ≥400 units/mg protein (Lowry)
Supelco
Dehydroepiandrosterone (DHEA) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
αα-Amilasa from Aspergillus oryzae, aqueous solution, ≥800 FAU/g
Sigma-Aldrich
α-Amylase from Bacillus amyloliquefaciens, liquid, ≥250 units/g
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~380 U/mg
Sigma-Aldrich
α-Amylase from porcine pancreas, Type I-A, PMSF treated, saline suspension, 700-1400 units/mg protein (E1%/280)
Supelco
αα-Amilasa from Bacillus licheniformis, suitable for determination of starch (Kit STA-20)