Merck
[Effect of L-lysine-a-oxidase on reproduction of herpes simplex type I virus in vitro].
Voprosy meditsinskoi khimii (1998-12-10)
I P Smirnova, S V Diorditsa, S B Alekseev, I Z Zaĭtsev
RESUMEN
L-lysin-a-oxidase (LO), a fungal enzyme catalysing oxidative deamination of L-lysin, was used for the inhibition of the reproduction of herpes simplex virus type 1 (HSV-1). Antiviral activity of LO was tested in vitro. The expression of viral antigens and CPE of HSV-1 was inhibited by LO at a concentration 0.7 mg/ml.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Lysine Oxidase from Trichoderma viride, lyophilized powder, ≥20 units/mg protein
En este momento no podemos mostrarle ni los precios ni la disponibilidad