Merck
Human monoamine oxidase A and B genes exhibit identical exon-intron organization.
Proceedings of the National Academy of Sciences of the United States of America (1991-05-01)
J Grimsby, K Chen, L J Wang, N C Lan, J C Shih
RESUMEN
Monoamine oxidases A and B [MAOA and MAOB; amine:oxygen oxidoreductase (deaminating) (flavin-containing), EC 1.4.3.4] play important roles in the metabolism of neuroactive, vasoactive amines and the Parkinsonism-producing neurotoxin 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP). Human MAOA and MAOB genes isolated from X chromosome-specific libraries span at least 60 kilobases, consist of 15 exons, and exhibit identical exon-intron organization. Exon 12 codes for the covalent FAD-binding-site and is the most conserved exon; the MAOA and MAOB exon 12 products share 93.9% peptide identity. These results suggest that MAOA and MAOB are derived from duplication of a common ancestral gene and provide insight on the structural/functional relationship of the enzyme products.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Monoamine Oxidase B human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Monoamine Oxidase A human, recombinant, expressed in baculovirus infected BTI insect cells