Merck
Solution structure and membrane interactions of the C2 domain of cytosolic phospholipase A2.
Journal of molecular biology (1998-07-17)
G Y Xu, T McDonagh, H A Yu, E A Nalefski, J D Clark, D A Cumming
RESUMEN
The amino-terminal, 138 amino acid C2 domain of cytosolic phospholipase A2 (cPLA2-C2) mediates an initial step in the production of lipid mediators of inflammation: the Ca2+-dependent translocation of the enzyme to intracellular membranes with subsequent liberation of arachidonic acid. The high resolution solution structure of this Ca2+-dependent, lipid-binding domain (CaLB) has been determined using heteronuclear three-dimensional NMR spectroscopy. Secondary structure analysis, derived from several sets of spectroscopic data, shows that the domain is composed of eight antiparallel beta-strands with six interconnecting loops that fits the "type II" topology for C2 domains. Using a total of 2370 distance and torsional restraints, the structure was found to be a beta-sandwich in the "Greek key" motif. The solution structure of cPLA2-C2 domain is very similar to the X-ray crystal structure of the C2 domain of phospholipase-C-delta and phylogenetic analysis clarifies the structural role of highly conserved residues. Calorimetric studies further demonstrate that cPLA2-C2 binds two Ca2+ with observed Kds of approximately 2 microM in an entropically assisted process. Moreover, regions on cPLA2-C2 interacting with membranes were identified by 15N-HSQC-spectroscopy of cPLA2-C2 in the presence of low molecular weight lipid micelles. An extended binding site was identified that binds the phosphocholine headgroup in a Ca2+-dependent manner and also interacts with proximal regions of the membrane surface. Based upon these results, a structural model is presented for the mechanism of association of cPLA2 with its membrane substrate.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Phospholipase A2 from honey bee venom (Apis mellifera), salt-free, lyophilized powder, 600-2400 units/mg protein
Sigma-Aldrich
Phospholipase A2 from porcine pancreas, ammonium sulfate suspension, ≥600 units/mg protein
Sigma-Aldrich
Phospholipase A2 from bovine pancreas, lyophilized powder, ≥20 units/mg protein