Merck
ppGpp accumulation in Pseudomonas aeruginosa and Pseudomonas fluorescens subjected to nutrient limitation and biocide exposure.
Letters in applied microbiology (2000-02-09)
D L Greenway, R R England
RESUMEN
The effect of a commonly used biocide, 1,2-benzisothiazolin-3-one (BIT) on ppGpp accumulation in the pathogen, Pseudomonas aeruginosa PAO1, and an environmental isolate, Ps. fluorescens, was examined. It is concluded that BIT is able to induce a stringent response in Ps. aeruginosa and Ps. fluorescens, determined by the rapid accumulation of ppGpp following addition of BIT to exponentially-growing cells. Western blot analysis of whole-cell extracts with anti-RelA antibody demonstrated that both species contain a RelA homologue. This is the first report of a RelA-like protein in pseudomonads.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
1,2-Benzisothiazol-3(2H)-one, 97%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
1,2-Benzisothiazol-3(2H)-one, analytical standard
En este momento no podemos mostrarle ni los precios ni la disponibilidad