- [Hydrolase from Xanthomonas rubrilineans for synthesis of cefalexin].
[Hydrolase from Xanthomonas rubrilineans for synthesis of cefalexin].
Prikladnaia biokhimiia i mikrobiologiia (2000-06-27)
A N Ivankin, A D Nekliudov
PMID10867949
RESUMEN
The use of peptide hydrolase (EC 3.4.13.1) from Xanthomonas rubrilineans for synthesis of the antibiotic cephalexin from 7-aminodesacetoxycephalosporanic acid was studied. The optimum conditions for production of cephalexin were determined, and the yield exceeded 80%. A method for monitoring the synthesis of this antibiotic synthesis by means of a conventional amino acid analyzer is proposed.
MATERIALES
Número de producto