Merck
Retrocorneal membrane after penetrating keratoplasty.
Ophthalmic surgery and lasers (2001-04-13)
T Lifshitz, T Oshry, G Rosenthal
RESUMEN
Retrocorneal membranes after penetrating keratoplasty (PKP) is a well known complications, resulting from unintentional retention of the host Descemet's membrane (DM), or donor DM detachment. We describe for the first time the formation of a retrocorneal inflammatory membrane that mimics donor DM detachment or retained recipient DM, and discuss the differential diagnosis of a retrocorneal membrane after PKP. Two patients who underwent PKP and subsequently developed a retrocorneal membrane are described. In both patients' eyes a retrocorneal membrane was observed immediately following penetrating keratoplasty. While the first patient's membrane persisted for 33 months in the presence of a clear graft, the second patient's membrane disintegrated and dissolved within two weeks following treatment with topical steroids. The first case describes a patient with retained Descemet's membrane, while in the second case, it turned out that the retrocorneal membrane was actually an inflammatory membrane.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Dexamethasone 21-acetate, ≥99%
Dexamethasone 21-acetate, European Pharmacopoeia (EP) Reference Standard