Merck
Monosaccharide esters: new tools in biomedicine.
Acta clinica Belgica (2002-08-03)
W J Malaisse
RESUMEN
The first report dealing with the use of monosaccharide esters as new tools in biomedicine was published in 1997 (1). This topic was first reviewed in 1998 (2). The major aim of the present report is to briefly evoke the background and present knowledge in this field.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
β-D-Glucose pentaacetate, 98%
Sigma-Aldrich
α-D(+)-Glucose pentaacetate, 99%