Merck
Effective recognition of H2PO4- by a new series of dendrimers functionalized with ferrocenyl-urea termini.
Chemical communications (Cambridge, England) (2002-08-29)
Beatriz Alonso, Carmen M Casado, Isabel Cuadrado, Moisés Morán, Angel E Kaifer
RESUMEN
A new series of dendrimers with poly(propylene imine) backbones and 4, 8, 16, or 32 peripheral ferrocenyl-urea groups were prepared and characterized; their voltammetric behavior in DMSO solution was very sensitive to the presence of hydrogenphosphate anions at submillimolar concentration levels.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Ferrocenecarboxylic acid, purum, ≥96%