Merck
  • Hypothalamic-pituitary-thyroid axis and sympathetic nervous system involvement in hyperthermia induced by 3,4-methylenedioxymethamphetamine (Ecstasy).
Hypothalamic-pituitary-thyroid axis and sympathetic nervous system involvement in hyperthermia induced by 3,4-methylenedioxymethamphetamine (Ecstasy).
The Journal of pharmacology and experimental therapeutics (2003-03-22)
Jon E Sprague, Matthew L Banks, Valerie J Cook, Edward M Mills
RESUMEN
An acute and potentially life-threatening complication associated with the recreational use of the 3,4-methylenedioxymethamphetamine (MDMA, Ecstasy) is hyperthermia. In the present study, Sprague-Dawley rats treated with MDMA (40 mg/kg s.c.) responded with a significant increase (maximal at 1 h) in rectal and skeletal muscle temperatures that lasted for at least 3 h post-treatment. Hypophysectomized (HYPO) and thyroparathyroidectomized (TX) animals treated with MDMA (40 mg/kg s.c.) did not become hyperthermic and in fact displayed a significant hypothermia. The HYPO and TX animals were also resistant to the serotonergic neurotoxic effects of MDMA assessed by serotonin measurements 4 to 7 days later in the striatum and hippocampus. MDMA (40 mg/kg s.c.) induced a significant increase in thyroxine levels 1 h post-treatment. Thyroid hormone replacement in TX animals returned the hyperthermic response seen after MDMA. Prazosin, an alpha(1)-antagonist (0.2 mg/kg i.p.), administered 30 min before MDMA significantly attenuated the MDMA-induced increase in rectal temperature, but had no effect on skeletal muscle temperature. Cyanopindolol, a beta(3)-antagonist (4 mg/kg s.c.), administered 30 min before MDMA (40 mg/kg s.c.) significantly attenuated the increase in skeletal muscle temperature, but had no effect on the rise in rectal temperature. The combination of prazosin and cyanopindolol resulted in an abolishment of MDMA-induced hyperthermia. The mechanisms of thermogenesis induced by MDMA seem to result from an interaction between the hypothalamic-pituitary-thyroid axis and the sympathetic nervous system, wherein mechanisms leading to core and skeletal muscle hyperthermia after MDMA exposure seem to be differentially regulated by alpha(1)- and beta(3)-adrenergic receptors.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
3,4-Methylenedioxyphenethylamine hydrochloride, 98%