Merck
Sequence encoding ribosomal protein L33 of Lactococcus lactis.
Gene (1992-09-21)
T Koivula, H Hemilä
RESUMEN
A cloned fragment from Lactococcus lactis chromosome encoding the L33 ribosomal protein was sequenced. Two incomplete open reading frames (ORFs) were also found: the upstream ORF shows similarity to the tetracycline-resistance protein (Tet) of Bacillus stearothermophilus, and the downstream ORF shows homology to a protein of Bacillus subtilis participating in sporulation (SpoVE), and to proteins of Escherichia coli involved in cell division (FtsW) and the maintenance of cell shape (RodA).
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
4-(Trifluoromethyl)aniline, 99%
Leflunomide impurity A, European Pharmacopoeia (EP) Reference Standard