Merck
  • Fluorogenic derivatization reagents suitable for isolation and identification of cysteine-containing proteins utilizing high-performance liquid chromatography-tandem mass spectrometry.
Fluorogenic derivatization reagents suitable for isolation and identification of cysteine-containing proteins utilizing high-performance liquid chromatography-tandem mass spectrometry.
Analytical chemistry (2004-01-31)
Mayumi Masuda, Chifuyu Toriumi, Tomofumi Santa, Kazuhiro Imai
RESUMEN
The fluorogenic derivatization reagents with a positive charge, 4-(dimethylaminoethylaminosulfonyl)-7-chloro-2,1,3-benzoxadiazole (DAABD-Cl) and 7-chloro-2,1,3-benzoxadiazole-4-sulfonylaminoethyltrimethylammonium chloride (TAABD-Cl), are proposed for use in proteomics studies. Following derivatization of protein mixtures with these reagents, a series of standard processes of isolation, digestion, and identification of the proteins were performed utilizing high-performance liquid chromatography-fluorescence detection and tandem mass spectrometry with the probability-based protein identification algorithm. Both DAABD and TAABD derivatives were detected fluorometrically at the femtomole level and showed more than 100-fold improvement in sensitivity compared to the underivatized original compounds with an electrospray ionization ion trap mass spectrometer analysis. The modification of the MASCOT database search system memorized with the fragment information of a DAABD-attached Cys residue allowed the identification of the proteolytic peptide fragments of the derivatized bovine serum albumin (BSA) with an estimated 38% sequence coverage of BSA. Utilizing DAABD-Cl as a derivatization reagent, identification of several proteins was also possible in a soluble extract of Caenorhabditis elegans (10 microg of protein). Consequently, for identification of proteins in the complex matrixes of proteins, DAABD-Cl could be a more appropriate reagent than ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate as reported previously.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
4-Fluoro-7-sulfamoylbenzofurazan