Merck
  • [Activity of NO-synthase, apoptosis signal-regulating protein kinase 1, poly-(ADP-ribose)-polymerase, protein kinase C, serine proteinases and DNA fragmentation in rat liver during molybdenum-dependent induction of xanthine oxidase ].
[Activity of NO-synthase, apoptosis signal-regulating protein kinase 1, poly-(ADP-ribose)-polymerase, protein kinase C, serine proteinases and DNA fragmentation in rat liver during molybdenum-dependent induction of xanthine oxidase ].
Ukrains'kyi biokhimichnyi zhurnal (1999 ) (2004-02-18)
V V Sumbaev
RESUMEN
It was discovered that nitric oxide synthase activity and concentration of S-nitrosothiols decreased during molybden-dependent xanthine oxidase activation. The increase of apoptosis signal-regulating kinase 1 activity, with the respected decrease of [table: see text] poly-(ADP-ribose)-polymerase and protein kinase C activities as well as increase of trypsine like serine proteinases activity were observed. No significant changes of DNA fragmentation were revealed.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium molybdate dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Sodium molybdate dihydrate, ≥99.5%
Sigma-Aldrich
Sodium molybdate, ≥98%
Sigma-Aldrich
Sodium molybdate dihydrate, ≥99.5%, suitable for plant cell culture
Sigma-Aldrich
Sodium molybdate, powder, −100 mesh particle size, 99.9% trace metals basis
Sigma-Aldrich
Sodium molybdate dihydrate, 99.99% trace metals basis