Merck
The use of novel water-in-oil microemulsions in microemulsion electrokinetic chromatography.
Electrophoresis (2004-02-26)
Kevin D Altria, Margo F Broderick, Sheila Donegan, Joe Power
RESUMEN
We describe the novel use of water-in-oil (W/O) microemulsions to achieve unique separations in microemulsion electrokinetic chromatography (MEEKC). The choice and concentration of the buffer type, surfactant and co-surfactant were all examined and optimized. Separations of a range of neutral and acidic analytes was shown to be markedly different to that obtained by (oil-in-water) O/W MEEKC. Neutral solutes are separated by virtue of their solubility (log P) values in O/W MEEKC with the more water-insoluble solutes migrating last. This separation process does not occur in W/O, as neutral solutes are not separated in order of log P.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
4′-Hydroxyacetophenone, 99%