Merck
Poly(dimethylsiloxane) electrospray devices fabricated with diamond-like carbon-poly(dimethylsiloxane) coated SU-8 masters.
Lab on a chip (2004-04-22)
K Huikko, P Ostman, K Grigoras, S Tuomikoski, V-M Tiainen, A Soininen, K Puolanne, A Manz, S Franssila, R Kostiainen, T Kotiaho
RESUMEN
This study presents coupling of a poly(dimethylsiloxane) (PDMS) micro-chip with electrospray ionization-mass spectrometry (ESI-MS). Stable electrospray is generated directly from a PDMS micro-channel without pressure assistance. Hydrophobic PDMS aids the formation of a small Taylor cone in the ESI process and facilitates straightforward and low-cost batch production of the ESI-MS chips. PDMS chips were replicated with masters fabricated from SU-8 negative photoresist. A novel coating, an amorphous diamond-like carbon-poly(dimethylsiloxane) hybrid, deposited on the masters by the filtered pulsed plasma arc discharge technique, improved significantly the lifetime of the masters in PDMS replications. PDMS chip fabrication conditions were observed to affect the amount of background peaks in the MS spectra. With an optimized fabrication process (PDMS curing agent/silicone elastomer base ratio of 1/8 (w/w), curing at 70 degree C for 48 h) low background spectra were recorded for the analytes. The performance of PDMS devices was examined in the ESI-MS analysis of some pharmaceutical compounds and amino acids.