Merck
Flow-induced molecular segregation in beta-casein-monoglyceride mixed films spread at the air-water interface.
Langmuir : the ACS journal of surfaces and colloids (2004-07-14)
Cecilio Carrera Sánchez, Juan M Rodríguez Patino
RESUMEN
In this work, we have used different and complementary interfacial techniques (surface film balance, Brewster angle microscopy, and interfacial shear rheology) to analyze the static (structure, topography, reflectivity, miscibility, and interactions) and flow characteristics (surface shear characteristics) of beta-casein and monoglyceride (monopalmitin and monoolein) mixed films spread on the air-water interface. The structural, topographical, and shear characteristics of the mixed films depend on the surface pressure and on the composition of the mixed film. The surface shear viscosity (etas) varies greatly with the surface pressure. In general, the greater the surface pressure, the greater the values of etas. At higher surface pressures, collapsed beta-casein residues may be displaced from the interface by monoglyceride molecules with important repercussions on the shear characteristics of the mixed films. A shear-induced change in the topography of monoglyceride and beta-casein domains, on one hand, and a segregation between domains of the film-forming components, on the other hand, were also observed. The displacement of the beta-casein by the monoglycerides is facilitated under shear conditions, especially for beta-casein-monoolein mixed films.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
DL-α-Palmitin, ≥99%