Merck
  • Chronoamperometry at micropipet electrodes for determination of diffusion coefficients and transferred charges at liquid/liquid interfaces.
Chronoamperometry at micropipet electrodes for determination of diffusion coefficients and transferred charges at liquid/liquid interfaces.
Analytical chemistry (2004-09-15)
Yi Yuan, Lei Wang, Shigeru Amemiya
RESUMEN
Chronoamperometry was carried out at liquid/liquid interfaces supported at the tip of micropipet electrodes for direct determination of the diffusion coefficient of a species in the outer solution. The diffusion coefficient was used for subsequent determination of the transferred charges per species from the diffusion-limited steady-state current. A large tip resistance of the micropipets causes prolonged charging current so that the faradic current can be measured accurately only at a long-time regime (typically t > 5 ms). At the same time, the long-time current response at the interfaces surrounded by a thin glass wall of the pipets is enhanced by diffusion of the species from behind the pipet tip. Therefore, numerical simulations of the long-time chronoamperometric response were carried out using the finite element method for accurate determination of diffusion coefficients. Validity of the simulation results was confirmed by studying simple transfer of tetraethylammonium ion. The technique was applied for transfer/adsorption reactions of the natural polypeptide protamine and also for Ca2+ and Mg2+ transfers facilitated by ionophore ETH 129. With the diffusion coefficient of protamine determined to be (1.2 +/- 0.1) x 10(-6) cm(2)/s, the ionic charge transferred by each protamine molecule was obtained as +20 +/- 1, which is close to the excess positive charge of protamine. Also, the diffusion coefficient of ETH 129 was determined to demonstrate that each ionophore molecule transfers +0.67 and +1 charge per Ca2+ and Mg2+ transfer, respectively, which corresponds to formation of 1:3 and 1:2 complexes with the respective ions.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
Calcium ionophore II, Selectophore, function tested