- Mapping the interaction of bradykinin 1-5 with the exodomain of human protease activated receptor 4.
Mapping the interaction of bradykinin 1-5 with the exodomain of human protease activated receptor 4.
FEBS letters (2004-12-29)
Marvin T Nieman, Eileen Pagan-Ramos, Mark Warnock, Yelena Krijanovski, Ahmed A K Hasan, Alvin H Schmaier
PMID15620686
RESUMEN
The angiotensin converting enzyme breakdown product of bradykinin, bradykinin 1-5 (RPPGF), inhibits thrombin-induced human or mouse platelet aggregation. RPPGF binds to the exodomain of human protease-activated receptor 1 (PAR1). Studies determined if RPPGF also binds to the exodomain of human PAR4. RPPGF binds to a peptide of the thrombin cleavage site on PAR4. Recombinant wild-type and mutated exodomain of human PAR4 was prepared. The N-terminal arginine on RPPGF binds to the P2 position or proline46 on PAR4 to block thrombin cleavage. These data indicate that RPPGF influences thrombin activity by binding to the thrombin cleavage site on both PAR4 and PAR1.
MATERIALES
Número de producto