Merck
Neonatal hyperammonemia: the N-carbamoyl-L-glutamic acid test.
The Journal of pediatrics (2005-08-30)
Nathalie Guffon, Manuel Schiff, David Cheillan, Bendicht Wermuth, Johannes Häberle, Christine Vianey-Saban
RESUMEN
In a prospective study, patients with a suspected urea cycle defect underwent oral N-carbamoyl-L-glutamic acid loading testing. In patients with subsequently confirmed N-acetylglutamate synthase deficiency, hyperammonemia normalized within 8 hours. This test may be useful in the early diagnosis of patients with suspected urea cycle disorders.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
N-Carbamyl-L-glutamic acid