Merck
Ligand-sensitized fluorescence of Tb3+ in Tb3+-dibutylphosphate complexes: application for the estimation of DBP.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2006-02-21)
S Maji, K S Viswanathan
RESUMEN
The fluorescence of Tb(3+) is sensitized by complexation with dibutylphosphate (DBP) and tri-n-butylphosphate (TBP). The excitation maximum for the Tb(3+)-DBP complex occurs at 218.5 nm, while that for the Tb(3+)-TBP complex is observed at 228.0 nm. Both complexes yield Tb(3+) fluorescence at 548 nm. The difference in the excitation maxima for the two complexes has been used to advantage for the estimation of DBP in the presence of TBP. DBP is the main degradation product of TBP in the PUREX process and the method described in this work can thus serve as a useful analytical tool in monitoring the quality of the TBP in the process. This method has been shown to be applicable for the estimation of DBP when present to an extent of 0.1-10% of TBP, in TBP/dodecane solutions.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Dibutyl phosphate, ≥97.0% (T)