Merck
Some aromatic hydrazone derivatives as inhibitors for the corrosion of C-steel in phosphoric acid solution.
Annali di chimica (2006-06-01)
Abd El-Aziz S Fouda, Ahmed A Al-Sarawy, Mohamed S Radwan
RESUMEN
The effect of furfural benzoylhydrazone and its derivatives (I-VII) as corrosion inhibitors for C-steel in 1M phosphoric acid solution has been studied by weight-loss and galvanostatic polarization techniques. A significant decrease in the corrosion rate of C-steel was observed in the presence of the investigated inhibitors. This study revealed that, the inhibition efficiency increases with increasing the inhibitor concentration, and the addition of iodide ions enhances it to a considerable extent. The effect of temperature on the inhibition efficiency of these compounds was studied using weight-loss method. Activation energy (E(a)*) and other thermodynamic parameters for the corrosion process were calculated and discussed. The galvanostatic polarization data indicated that, the inhibitors were of mixed-type, but the cathode is more polarized than the anode. The adsorption of these compounds on C-steel surface has been found to obey Frumkin's adsorption isotherm. The mechanism of inhibition was discussed in the light of the chemical structure of the undertaken inhibitors.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Benzhydrazide, 98%