Merck
Effects of isopropyl palmitate on the skin permeation of drugs.
Biological & pharmaceutical bulletin (2006-11-02)
Hong Guo, Zhidong Liu, Jizhong Li, Shufang Nie, Weisan Pan
RESUMEN
The model penetrants oxaprozin, nimesulide, gliclazide, and ribavirin, because of their different lipophilicities, were selected to assess the enhancing activity of pre-treatment solutions consisting of isopropyl palmitate (IP) in ethanol (5%, 10%, 15%and 20%, w/w, respectively) across excised rat skin using Franz diffusion cells and HPLC detection. All pre-treatment solutions produced a significant increase in the flux and permeation of all four penetrants (p<0.001) and a relationship between penetrant lipophilicity and enhancement effect was observed. The general order of IP effectiveness at concentration was 20%>15%>10%>5% (w/w). The lag-time of drugs did not significantly change except for ribavirin.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Isopropyl palmitate, technical grade, 90%
Sigma-Aldrich
Isopropyl palmitate, ≥90%
Supelco
Isopropyl palmitate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Oxaprozin, solid