Merck
Effect of clomazone herbicide on biochemical and histological aspects of silver catfish (Rhamdia quelen) and recovery pattern.
Chemosphere (2007-02-07)
Márcia Crestani, Charlene Menezes, Lissandra Glusczak, Denise dos Santos Miron, Roselia Spanevello, Aron Silveira, Fábio Ferreira Gonçalves, Renato Zanella, Vânia Lúcia Loro
RESUMEN
The effects of the herbicide, clomazone, on acetylcholinesterase (AChE), catalase and TBARS formation in teleost fish (Rhamdia quelen) were studied. The fish were exposed to 0.5 or 1.0 mg L(-1) of clomazone for 12, 24, 48, 96 and 192 h. After 192 h of exposure period, fish were transferred to clean water and kept in the same for 192 h to study the recovery response. Same parameters as that of exposure period were assayed after 96 and 192 h of recovery period. Specific AChE activity was reduced in the brain and muscle after treatments, reaching a maximum inhibition of 47% in the brain and 45% in the muscle after 12h of exposure. Fish exposed to clomazone increased TBARS production in the liver for all exposure periods. The brain presented elevated TBARS levels after 12, 24 and 48 h, but after 96 and 192 h, these levels decreased. The decrease of TBARS levels persisted in brain tissue after 96 h of recovery and returned to the control value after 192 h in clean water. Catalase activity was reduced for all periods of exposure. Histological analysis showed vacuolation in the liver after herbicide exposure. Some of the alterations observed were completely restored after recovery period.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
Clomazone, PESTANAL®, analytical standard