Merck
Direct oxidation of n-heptane to ester over modified sulfated SnO2 catalysts under mild conditions.
Journal of hazardous materials (2007-02-27)
Xue Cui, Hongzhu Ma, Bo Wang, Hongwei Chen
RESUMEN
The direct oxidation of n-heptane to ester using air as the oxidant under mild conditions assisted by sulfated tin oxide (SO(4)(2-)/SnO(2)) and Co modified sulfated tin oxide (SO(4)(2-)/SnO(2)-Co(2)O(3)) prepared by chemical co-precipitation method and characterized by means of SEM, XRD, FT-IR and XPS techniques was studied. From the results it was found that the catalytic activity of SO(4)(2-)/SnO(2)-Co(2)O(3) is higher than that of SO(4)(2-)/SnO(2) in this target reaction, and two asymmetric esters: diisobutyl phthalate and cyclohexylmethyl tridecyl oxalate that have not been reported in the directly oxidation of n-heptane by traditional method have been obtained. The possible mechanism was also discussed.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Diisobutyl phthalate, 99%
Supelco
Diisobutyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland