Merck
Tailor-made fructooligosaccharides by a combination of substrate and genetic engineering.
Chembiochem : a European journal of chemical biology (2007-12-07)
Alga Zuccaro, Sven Götze, Susanne Kneip, Petra Dersch, Jürgen Seibel
RESUMEN
The combination of sucrose analogues as novel substrates (substrate engineering) and highly active recombinant beta-fructofuranosidase from A. niger (genetic engineering) provides a new powerful tool for the efficient preparative synthesis of tailor-made saccharides of the important 1-kestose and 1-nystose type headed with different monosaccharides of interest. These novel compounds have been isolated. They did not display toxic effects or suppress cell growth in initial studies, making these new compounds potential candidates for prebiotics.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
1-Kestose, analytical standard