Merck
Heat-set gels and egg-like vesicles using two component gel system based on chiral calix[4]arenes.
Chemical communications (Cambridge, England) (2007-12-07)
Jin-Lan Zhou, Xian-Jie Chen, Yan-Song Zheng
RESUMEN
Chiral calix[4]arenes bearing long tertiary alkyl groups at the upper rim and S-1-phenylethylamine groups at the lower rim can form heat-set gels and egg-like vesicles enantioselectively with d-2,3-dibenzoyltartaric acid in cyclohexane, which is the first example of heat-set gels resulting from difference in interactions between two component gelators: in addition, the diameter of vesicles decreased with the increase in length of alkyl groups, which could be used to control the size of the vesicles.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Dibenzoyl-L-tartaric acid, 98%
Sigma-Aldrich
(+)-2,3-Dibenzoyl-D-tartaric acid, ≥98%, made from synthetic tartaric acid
Sigma-Aldrich
(+)-2,3-Dibenzoyl-D-tartaric acid, ≥99.0% (T)