Merck
5-Hydroxytryptamine2A receptor binding activity of compounds from Litsea sessilis.
Phytotherapy research : PTR (2008-10-11)
Lip Yong Chung, Mee Wah Lo, Muhammad Rais Mustafa, Swee Hock Goh, Zamrie Imiyabir
RESUMEN
A 96-well microplate filtration based 5-HT(2A) receptor-radioligand binding assay was optimized and adopted to carry out a bioassay-guided fractionation of the methanol extract of the leaves of Litsea sessilis. This purification led to the isolation of two compounds identified as (+)-boldine (1) and (+)-dehydrovomifoliol (2). (+)-Boldine binds to 5-HT(2A) receptors at high concentrations with a K(i) value of 2.16 microm. However, (+)-dehydrovomifoliol showed minimal competitive inhibition on the binding of [(3)H]ketanserin to the same receptor with a K(i) value of 2.06 mm. These results suggest that (+)-boldine influences the activity of 5-HT(2A) receptors through competitive binding as an agonist or antagonist.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
Boldine, analytical standard