Merck
Spectroscopic evidence for an oxazolone structure of the b(2) fragment ion from protonated tri-alanine.
Journal of the American Society for Mass Spectrometry (2008-11-18)
Jos Oomens, Sarah Young, Sam Molesworth, Michael van Stipdonk
RESUMEN
Infrared multiple photon dissociation (IRMPD) spectroscopy is used to identify the structure of the b(2)(+) ion generated from protonated tri-alanine by collision induced dissociation (CID). The IRMPD spectrum of b(2)(+) differs markedly from that of protonated cyclo-alanine-alanine, demonstrating that the product is not a diketopiperazine. Instead, comparison of the IRMPD spectrum of b(2)(+) to spectra predicted by density functional theory provides compelling evidence for an oxazolone structure protonated at the oxazolone N-atom.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Ala-Ala-Ala