Merck
  • Simultaneous measurement of diazolidinyl urea, urea, and allantoin in cosmetic samples by hydrophilic interaction chromatography.
Simultaneous measurement of diazolidinyl urea, urea, and allantoin in cosmetic samples by hydrophilic interaction chromatography.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2009-03-10)
Takahiro Doi, Keiji Kajimura, Satoshi Takatori, Naoki Fukui, Shuzo Taguchi, Shozo Iwagami
RESUMEN
A new HPLC method for simultaneous measurement of diazolidinyl urea (DU), urea, and allantoin by hydrophilic interaction chromatography using a column packed with triazol-bonded silica particles is described. The calibration curves of DU, urea, and allantoin were linear over the ranges 2.5-125.0, 30-1250, and 0.25-18.75mg/L, respectively. The recoveries of DU, urea, and allantoin from homemade cosmetic samples ranged from 92.84% to 101.69%, 98.19% to 103.22%, and 95.75% to 102.09%, respectively. The peak relative standard deviation (RSD) values for the recovery tests of 3 concentrations/5 replicates were 3.03% for all compounds. In day-to-day measurement of recovery tests from homemade lotions over 3 consecutive days, the RSDs were less than 2.5% in all cases. This method was well validated and would be helpful for quickly analyzing these compounds in cosmetic samples.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Diazolidinyl urea, ≥95%