Merck
  • Investigating effects of bromide ions on trihalomethanes and developing model for predicting bromodichloromethane in drinking water.
Investigating effects of bromide ions on trihalomethanes and developing model for predicting bromodichloromethane in drinking water.
Water research (2010-01-19)
Shakhawat Chowdhury, Pascale Champagne, P James McLellan
RESUMEN
Chlorination for drinking water can form brominated trihalomethanes (THMs) in the presence of bromide ions. Recent studies have reported that bromodichloromethane (BDCM) has a stronger association with stillbirths and neural tube defects than other THMs species. In this paper, the results of an experimental investigation into the factors forming THMs in the presence of bromide ions are presented. The experiments were conducted using synthetic water samples with different characteristics (e.g., pH, temperature, dissolve organic content). Different combinations of these characteristics were considered in the experimental program. The results showed that increased bromide ion concentrations led to increases in the formation of total THMs, with higher BDCM and dibromochloromethane (DBCM), and lower chloroform formation. By increasing the pH from 6 to 8.5, increased chloroform and decreased BDCM and DBCM formation were observed. Higher bromide ions to chlorine ratios increased BDCM and DBCM and decreased chloroform formation, while higher temperatures increased BDCM, DBCM and chloroform formation. In most cases, bromoform (CHBr(3)) concentrations were found to be below the detection limit. Significant factors influencing BDCM formation were identified using a statistical analysis. A model for BDCM formation was estimated from 44 experiments and statistical adequacy was assessed using appropriate diagnostics, including residual plots and an R(2) of 0.97. The model was validated using external data from 17 water supply systems in Newfoundland, Canada. The predictive performance of the model was found to be excellent, and the resulting model could be used to predict BDCM formation in drinking water and to perform risk-cost balance analyses for best management practices.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Bromodichloromethane, ≥97%
Sigma-Aldrich
Dibromochloromethane, 97%