Merck
Interaction of flavonoids, the naturally occurring antioxidants with different media: a UV-visible spectroscopic study.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2010-02-19)
Bushra Naseem, S W H Shah, Aurangzeb Hasan, S Sakhawat Shah
RESUMEN
Quantitative parameters for interaction of flavonoids-the naturally occurring antioxidants, with solvents and surfactants are determined using UV-visible absorption spectroscopy. The availability of flavonoids; kaempferol, apigenin, kaempferide and rhamnetin in micelles of sodium dodecyl sulfate (SDS) is reflected in terms of partition coefficient, K(c). Thermodynamic calculations show that the process of transfer of flavonoid molecules to anionic micelles of SDS is energy efficient. A distortion in flavonoid's morphology occurs in case of kaempferol and apigenin in surfactant and water, exhibited in terms of a new band in the UV region of electronic spectra of these flavonoids. The partition coefficients of structurally related flavonoids are correlated with their antioxidant activities.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
Rhamnetin, analytical standard