Merck
  • Effects of cholesterol and nuclear hormone receptor agonists on the production of transforming growth factor-beta in macrophages.
Effects of cholesterol and nuclear hormone receptor agonists on the production of transforming growth factor-beta in macrophages.
Bulletin of experimental biology and medicine (2010-04-17)
Ya Sh Schwartz, O M Khoshchenko, M I Dushkin, N A Feofanova
RESUMEN
We studied the effects of cholesterol, its oxidized derivatives mevalonate, and nuclear receptor agonists LXR, RXR, and FXR on the production of transforming growth factor-beta1 (TGF- beta1) by macrophages. After recruiting of macrophage monocytes into the focus of inflammation, the production of TGF-beta1 increased by 3.5 times in comparison with control macrophages. Cholesterol diet stimulated the production of TGF-beta1 by 2.5 times. Cholesterol directly stimulated macrophage production of TGF-beta1 in vitro, while addition of mevalonate to the incubation medium effectively reduced this induced production. Agonists of nuclear receptor sharply reduced the production of TGF-beta1 in recruited macrophages. Under conditions of inflammation, hypercholesterolemia can be a factor of fibrogenesis due to TGF-beta1 induction in macrophages, which depends on the products of mevalonate biochemical chain.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
(±)-Mevalonolactone, ~97% (titration)