Merck
The role of tryptophan catabolism along the kynurenine pathway in acute ischemic stroke.
Neurochemical research (2010-05-22)
Raf Brouns, Robert Verkerk, Tony Aerts, Didier De Surgeloose, Annick Wauters, Simon Scharpé, Peter P De Deyn
RESUMEN
Post-stroke inflammation may induce upregulation of the kynurenine (KYN) pathway for tryptophan (TRP) oxidation, resulting in neuroprotective (kynurenic acid, KA) and neurotoxic metabolites (3-hydroxyanthranillic acid, 3-HAA). We investigated whether activity of the kynurenine pathway in acute ischemic stroke is related to initial stroke severity, long-term stroke outcome and the ischemia-induced inflammatory response. Plasma concentrations of TRP and its metabolites were measured in 149 stroke patients at admission, at 24 h, at 72 h and at day 7 after stroke onset. We evaluated the relation between the KYN/TRP ratio, the KA/3-HAA ratio and stroke severity, outcome and inflammatory parameters (C-reactive protein (CRP), erythrocyte sedimentation rate (ESR) and neutrophil/lymphocyte ratio (NLR)). KYN/TRP but not KA/3-HAA correlated with the NIHSS score and with the infarct volume. Patients with poor outcome had higher mean KYN/TRP ratios than patients with more favourable outcome. The KYN/TRP ratio at admission correlated with CRP levels, ESR and NLR. The activity of the kynurenine pathway for tryptophan degradation in acute ischemic stroke correlates with stroke severity and long-term stroke outcome. Tryptophan oxidation is related to the stroke-induced inflammatory response.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
3-Hydroxyanthranilic acid, 97%