Merck
  • Simultaneous voltammetric determination of ascorbic acid, dopamine and uric acid using polybromothymol blue film-modified glassy carbon electrode.
Simultaneous voltammetric determination of ascorbic acid, dopamine and uric acid using polybromothymol blue film-modified glassy carbon electrode.
Chemical & pharmaceutical bulletin (2010-06-05)
Xiongwei Xu, Qihuang Lin, Ailin Liu, Wei Chen, Xiuhua Weng, Changlian Wang, Xinhua Lin
RESUMEN
A sensitive and selective electrochemical method for simultaneous determination of ascorbic acid (AA), dopamine (DA), and uric acid (UA) using an electropolymerized bromothymol blue (BTB)-modified glassy carbon electrode (GCE) was developed. The electrochemically synthesized film was investigated using electrochemical impedance spectroscopy and voltammetric methods. The electrochemical behavior of the polymer-modified electrode depends on film thickness, i.e., the electropolymyerization time. The poly-BTB-modified GCE shows excellent electrocatalytic activity toward the oxidation of AA, DA, and UA in phosphate buffer solution (pH 5.0). The voltametric peak separations of AA/DA, DA/UA, and AA/UA on this modified electrode are 118 mV, 298 mV, and 455 mV, respectively. Therefore the voltammetric responses of these three compounds can be resolved well on the polymer-modified electrode, and simultaneous determination of these three compounds can be achieved. In addition, this modified electrode can be successfully applied to determine AA and DA in injection and UA in urine samples without interference.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Bromothymol Blue, ACS reagent, Dye content 95 %