Merck
  • An improved method for analyzing chlormequat and mepiquat in source waters by solid-phase extraction and liquid chromatography-mass spectrometry.
An improved method for analyzing chlormequat and mepiquat in source waters by solid-phase extraction and liquid chromatography-mass spectrometry.
Analytica chimica acta (2010-09-28)
Zhiyong Yu, Fen Jin, Jianying Hu, Xi Zhang, Jingfang Sun, Min Yang
RESUMEN
A liquid chromatography-mass spectrometry (LC-MS) method for the identification and quantification of chlormequat (CQ) and mepiquat (MQ) in source waters with high sensitivity and specificity was established using WCX cartridges (150 mg/6 mL) for pre-concentration of the samples and using the CAPCELL PAK CR 1:4 (2.0 mm×150 mm 5 μm, SCX:C18=1:4) column containing strong cationic exchange resins and C18 for separation. The method could solve the problem for pre-concentrating ionic compounds from water samples and avoid the MS instrument fouling problem accompanied with the use of ion-pair reagents. After the optimization of analytical conditions, quantification was achieved in the positive electrospray ionization mode using selected ion monitoring. The samples were analyzed with multi-channel mode with quantification performed at 30 V cone voltage to ascertain the sensitivity and qualitative analysis performed at 60 V cone voltage simultaneously. The method detection limits (MDLs) of CQ and MQ in source water were determined to be 14 and 22 ng L(-1). Finally, the method was used to quantify CQ and MQ in several source waters, which gave a level ranging from below the quantitation limit to 28 ng L(-1).
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
(2-Chloroethyl)trimethylammonium chloride, 98%
Sigma-Aldrich
Chlorocholine chloride, BioReagent, suitable for plant cell culture
Supelco
Mepiquat chloride, PESTANAL®, analytical standard
Supelco
Chlormequat chloride, PESTANAL®, analytical standard