Merck
  • Otoacoustic emissions in bushcricket ears: general characteristics and the influence of the neuroactive insecticide pymetrozine.
Otoacoustic emissions in bushcricket ears: general characteristics and the influence of the neuroactive insecticide pymetrozine.
Journal of comparative physiology. A, Neuroethology, sensory, neural, and behavioral physiology (2010-11-06)
Doreen Möckel, Ernst-August Seyfarth, Manfred Kössl
RESUMEN
The tympanal organ of the bushcricket Mecopoda elongata emits pronounced distortion-product otoacoustic emissions (DPOAEs). Their characteristics are comparable to those measured in other insects, such as locusts and moths, with the 2f1-f2 emission being the most prominent one. Yet the site of their generation is still unclear. The spatial separation between the sound receiving spiracle and the hearing organ in this species allows manipulations of the sensory cells without interfering with the acoustical measurements. We tried to interfere with the DPOAE generation by pharmacologically influencing the tympanal organ using the insecticide pymetrozine. The compound appears to act selectively on scolopidia, i.e., the mechanosensor type characteristically constituting tympanal organs. Pymetrozine solutions were applied as closely as possible to the scolopidia via a cuticle opening in the tibia, distally to the organ. Applications of pymetrozine at concentrations between 10(-3) and 10(-7) M to the tympanal organ led to a pronounced and irreversible decrease of the DPOAE amplitudes.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
Pymetrozin, PESTANAL®, analytical standard