Merck
Desymmetrization of cyclohexadienones via asymmetric Michael reaction catalyzed by cinchonine-derived urea.
Organic letters (2011-09-15)
Qing Gu, Shu-Li You
RESUMEN
Desymmetrization of cyclohexadienones bearing a bisphenylsulfonyl methylene group via asymmetric Michael reaction catalyzed by cinchonine-derived urea was realized to afford a series of highly enantioenriched polycyclic cyclohexenones in high yields and ee's.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
(+)-Cinchonine, 85%
Sigma-Aldrich
Cinchonine, crystallized, ≥98.0% (NT)